Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175375.3 | NP_780584.2 | 27 | 5099 | ||
GenBank mRNA | BC091647.1 | - | 3 | 524 |