Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aif1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145144.1 | NP_660126.1 | 5, 6 | 420 | ||
GenBank mRNA | AB035322.1 | - | 5, 6 | 372 | ||
AK028955.1 | - | 5, 6 | 378 | |||
AK045539.1 | - | 5, 6 | 450 | |||
BC024599.1 | - | 5, 6 | 420 | 5044495 |