Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sccpdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178653.3 | NP_848768.1 | 1 | 149 | ||
GenBank mRNA | AK042919.1 | - | 1 | 37 | ||
AK046033.1 | - | 1 | 150 | |||
AK046468.1 | - | 1 | 136 | |||
AK050554.1 | - | 1 | 96 | |||
AK077695.1 | - | 1 | 99 | |||
AK081021.1 | - | 1 | 121 | |||
AK082803.1 | - | 1 | 34 | |||
AK086448.1 | - | 1 | 171 | |||
AK086632.1 | - | 1 | 147 | |||
AK145399.1 | - | 1 | 133 | |||
AK166018.1 | - | 1 | 61 | |||
BC025803.1 | - | 1 | 146 | 5100552 |