Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sccpdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178653.3 | NP_848768.1 | 2 | 406 | ||
GenBank mRNA | AK042919.1 | - | 2 | 294 | ||
AK046033.1 | - | 2 | 407 | |||
AK046468.1 | - | 2 | 393 | |||
AK077695.1 | - | 2 | 356 | |||
AK081021.1 | - | 1 | 2415 | |||
AK082803.1 | - | 2 | 291 | |||
AK086448.1 | - | 2 | 429 | |||
AK086632.1 | - | 2 | 404 | |||
AK145399.1 | - | 2 | 390 | |||
AK166018.1 | - | 2 | 318 | |||
AK182882.1 | - | 2 | 148 | |||
AK215316.1 | - | 2 | 148 | |||
BC025803.1 | - | 2 | 403 | 5100552 |