Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027321.3 | NP_081597.1 | 4 | 555 | ||
NM_175413.3 | NP_780622.1 | 4 | 555 | |||
GenBank mRNA | AK008119.1 | - | 4 | 523 | ||
AK035072.1 | - | 4 | 557 | |||
AK039277.1 | - | 4 | 552 | |||
AK046671.1 | - | 4 | 451 | |||
AK052159.1 | - | 4 | 507 | |||
AK079125.1 | - | 4 | 533 | |||
BC119529.2 | - | 4 | 504 | 40056120 | ||
BC119530.1 | - | 4 | 505 | 40056121 |