Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3hdm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177994.4 | NP_818775.2 | 3, 4 | 385 | ||
GenBank mRNA | AK081288.1 | - | 3, 4 | 374 | ||
AK088107.1 | - | 3, 4 | 386 | |||
AK139047.1 | - | 3, 4 | 376 | |||
AK159869.1 | - | 3, 4 | 364 | |||
AK165610.1 | - | 3, 4 | 356 | |||
BC034653.1 | - | 2, 3 | 188 | |||
BC095970.1 | - | 3, 4 | 366 | 6530790 | ||
BC132463.1 | - | 3, 4 | 461 | 40130740 | ||
BC138461.1 | - | 3, 4 | 461 | 8861481 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK048569.1 | - | 577 | Chr. - - [-] on Build |