Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025469.2 | NP_079745.1 | 1, 2 | 155 | ||
GenBank mRNA | AF414676.1 | - | 1, 2 | 142 | ||
AF414677.1 | - | 1, 2 | 142 | |||
AK008635.1 | - | 1, 2 | 153 | |||
AK008815.1 | - | 1, 2 | 156 | |||
AK008834.1 | - | 1, 2 | 153 | |||
AK008839.1 | - | 1, 2 | 153 | |||
AK008874.1 | - | 1, 2 | 153 | |||
AK008879.1 | - | 1, 2 | 152 | |||
AK019005.1 | - | 1, 2 | 153 | |||
AK019047.1 | - | 1, 2 | 153 | |||
AK028141.1 | - | 1, 2 | 155 | |||
BC042935.1 | - | 1, 2 | 140 | 6399295 |