Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148934.2 | NP_683736.1 | 1 | 139 | ||
GenBank mRNA | AF303106.1 | - | 1 | 139 | ||
AK153467.1 | - | 1 | 66 | |||
BC138515.1 | - | 1 | 180 | 8861535 | ||
BC138516.1 | - | 1 | 180 | 8861536 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK077345.1 | - | 671 | Chr. - - [-] on Build |