Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025900.2 | NP_080176.2 | 7 | 830 | ||
GenBank mRNA | AK007546.1 | - | 4 | 379 | ||
AK012615.1 | - | 7 | 771 | |||
AK012877.1 | - | 7 | 765 | |||
AK033451.1 | - | 7 | 720 | |||
AK141790.1 | - | 1 | 182 | |||
BC048844.1 | - | 7 | 780 | |||
BC055451.1 | - | 7 | 776 | 4164972 |