Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286573.1 | NP_001273502.1 | 18 | 2559 | ||
NM_001286575.1 | NP_001273504.1 | 20 | 2497 | |||
NM_145830.2 | NP_665829.1 | 19 | 2661 | |||
NM_147151.2 | NP_671493.1 | 19 | 2395 | |||
GenBank mRNA | AB077209.1 | - | 19 | 2388 | ||
AB077210.1 | - | 19 | 2661 | |||
AK077743.1 | - | 7 | 717 | |||
AK154246.1 | - | 18 | 2582 | |||
AK182117.1 | - | 2 | 84 | |||
BC058357.1 | - | 20 | 2495 | |||
BC096670.1 | - | 5 | 455 |