Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb2b26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178308.2 | NP_840093.1 | 3 | 284 | ||
GenBank mRNA | AY293281.1 | - | 3 | 280 | ||
AY293282.1 | - | 2 | 194 | |||
BC035222.1 | - | 3 | 270 | 4192901 | ||
BC107292.1 | - | 3 | 272 | 40053055 | ||
BC107293.1 | - | 3 | 272 | 40053056 | ||
BC111446.1 | - | 3 | 284 | 40053210 | ||
BC134362.1 | - | 3 | 290 | 40134659 | ||
BC148302.2 | - | 3 | 290 | 40134662 |