Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081274.1 | NP_001074743.1 | 10 | 1130 | ||
GenBank mRNA | AK002894.1 | - | 10 | 1092 | ||
AK004217.1 | - | 3 | 334 | |||
AK077110.1 | - | 10 | 1089 | |||
AK145602.1 | - | 10 | 1094 | |||
AK150210.1 | - | 10 | 1093 | |||
AK150490.1 | - | 10 | 1089 | |||
AK151380.1 | - | 10 | 1089 | |||
AK151993.1 | - | 10 | 1089 | |||
AK152758.1 | - | 10 | 1089 | |||
AK152894.1 | - | 10 | 1089 | |||
AK153409.1 | - | 10 | 1086 | |||
AK155027.1 | - | 10 | 1089 | |||
AK166733.1 | - | 10 | 1084 | |||
AK166947.1 | - | 10 | 1066 | |||
AK167215.1 | - | 10 | 1084 | |||
AK168251.1 | - | 10 | 1089 | |||
AK184317.1 | - | 2 | 129 | |||
AK217709.1 | - | 2 | 129 | |||
BC008646.1 | - | 10 | 1061 | 3155147 | ||
BC011329.1 | - | 10 | 1079 | 3984654 |