Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Triobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039155.1 | NP_001034244.1 | 8 | 2431 | ||
NM_001039156.1 | NP_001034245.1 | 8 | 2431 | |||
NM_138579.4 | NP_613045.3 | 8 | 2431 | |||
GenBank mRNA | AK155197.1 | - | 1 | 134 | ||
BC094366.1 | - | 1 | 1088 | |||
DQ228000.1 | - | 8 | 2431 | |||
DQ228001.2 | - | 8 | 2431 | |||
DQ228002.2 | - | 8 | 2431 | |||
DQ278602.1 | - | 7 | 2050 |