Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025628.2 | NP_079904.1 | 2, 3 | 213 | ||
GenBank mRNA | AK007996.1 | - | 2, 3 | 214 | ||
AK012686.1 | - | 2, 3 | 179 | |||
AK018721.1 | - | 2, 3 | 148 | |||
BC024343.1 | - | 2, 3 | 156 | 4989436 |