Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290818.1 | NP_001277747.1 | 8 | 781 | ||
NM_130863.2 | NP_570933.1 | 9 | 853 | |||
GenBank mRNA | AF333028.1 | - | 8 | 573 | ||
AK155691.1 | - | 8 | 782 | |||
AK193363.1 | - | 3 | 94 | |||
BC003196.1 | - | 8 | 569 | |||
BC033272.1 | - | 7 | 897 | |||
BC053922.1 | - | 9 | 586 | 6309555 |