Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290684.1 | NP_001277613.1 | 4 | 1389 | ||
NM_172441.3 | NP_766029.2 | 4 | 1389 | |||
GenBank mRNA | DQ358971.1 | - | 4 | 1370 | ||
EF071946.1 | - | 4 | 1389 |