Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144784.3 | NP_659033.1 | 3 | 250 | ||
GenBank mRNA | AJ510150.1 | - | 3 | 242 | ||
AK032097.1 | - | 3 | 221 | |||
AK075593.1 | - | 3 | 220 | |||
AK081715.1 | - | 3 | 250 | |||
AK146241.1 | - | 3 | 205 | |||
AK163402.1 | - | 3 | 254 | |||
AK169771.1 | - | 3 | 243 | |||
BC024429.1 | - | 3 | 253 | |||
BC024763.1 | - | 3 | 222 | 5365469 |
Gene Symbol: Fn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC085240.1 | - | 208 |