Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083906.1 | NP_001077375.1 | 7 | 2547 | ||
GenBank mRNA | AK136580.1 | - | 4 | 590 | ||
BC133713.1 | - | 7 | 2547 | 40143429 |