Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145129.2 | NP_660111.2 | 2 | 340 | ||
GenBank mRNA | AF325346.1 | - | 2 | 210 | ||
AF459029.1 | - | 2 | 313 | |||
AF472588.1 | - | 2 | 242 | |||
AK051730.1 | - | 2 | 344 | |||
AK053497.1 | - | 2 | 268 | |||
AK080415.1 | - | 2 | 341 | |||
AK080475.1 | - | 2 | 341 | |||
AK148786.1 | - | 2 | 312 | |||
AY574262.1 | - | 2 | 175 | |||
BC120597.1 | - | 2 | 211 | 40129520 | ||
BC120599.1 | - | 2 | 211 | 40129522 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK053712.1 | - | 2114 | Chr. - - [-] on Build |