Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145129.2 | NP_660111.2 | 5 | 833 | ||
GenBank mRNA | AF325346.1 | - | 5 | 703 | ||
AF459029.1 | - | 5 | 806 | |||
AF472588.1 | - | 5 | 735 | |||
AK051730.1 | - | 5 | 837 | |||
AK053497.1 | - | 5 | 761 | |||
AK080415.1 | - | 5 | 834 | |||
AK080475.1 | - | 5 | 836 | |||
AK148786.1 | - | 5 | 805 | |||
AY574262.1 | - | 5 | 668 | |||
BC120597.1 | - | 5 | 704 | 40129520 | ||
BC120599.1 | - | 5 | 704 | 40129522 |