Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabra5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176942.4 | NP_795916.1 | 6, 7 | 930 | ||
GenBank mRNA | AK038476.1 | - | 6, 7 | 932 | ||
AK083185.1 | - | 6, 7 | 717 | |||
BC062112.1 | - | 6, 7 | 928 | 6815819 |