Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144803.2 | NP_659052.1 | 5 | 427 | ||
GenBank mRNA | AY574261.1 | - | 4 | 311 | ||
BC011490.1 | - | 5 | 427 | 4193582 |