Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077190.2 | NP_001070658.1 | 2 | 386 | ||
NM_001077192.2 | NP_001070660.1 | 2 | 386 | |||
NM_001077193.2 | NP_001070661.1 | 2 | 386 | |||
NM_007380.3 | NP_031406.2 | 2 | 386 | |||
NM_145994.2 | NP_666106.1 | 2 | 386 | |||
GenBank mRNA | AF420251.1 | - | 2 | 322 | ||
AK034476.1 | - | 2 | 268 | |||
AK151026.1 | - | 2 | 294 | |||
AK151363.1 | - | 2 | 365 | |||
AK152061.1 | - | 2 | 250 | |||
AK152184.1 | - | 2 | 250 | |||
AK153130.1 | - | 2 | 294 | |||
AK164040.1 | - | 2 | 302 | |||
AK165924.1 | - | 2 | 359 | |||
AK167328.1 | - | 2 | 345 | |||
AK167568.1 | - | 2 | 341 | |||
AK167744.1 | - | 2 | 333 | |||
AY033645.1 | - | 2 | 230 | |||
BC004657.1 | - | 2 | 337 | 3498068 | ||
BC079642.1 | - | 2 | 292 | 6816252 | ||
BY278454.1 | - | 2 | 386 |