Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114595.1 | NP_001108067.1 | 13, 14 | 2067 | ||
NM_001114596.1 | NP_001108068.1 | 14, 15 | 2103 | |||
NM_001114597.1 | NP_001108069.1 | 14, 15 | 4935 | |||
NM_053252.3 | NP_444482.2 | 14, 15 | 2412 | |||
GenBank mRNA | AK087817.1 | - | 10, 11 | 1654 |