Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114595.1 | NP_001108067.1 | 3, 4 | 555 | ||
NM_001114596.1 | NP_001108068.1 | 3, 4 | 555 | |||
NM_001114597.1 | NP_001108069.1 | 3, 4 | 555 | |||
NM_053252.3 | NP_444482.2 | 3, 4 | 555 | |||
GenBank mRNA | AF305087.1 | - | 3, 4 | 508 | ||
AF305088.1 | - | 3, 4 | 508 | |||
AF305089.1 | - | 3, 4 | 508 | |||
AF305090.1 | - | 3, 4 | 508 | |||
AK089917.1 | - | 3, 4 | 516 | |||
AK169974.1 | - | 3, 4 | 521 | |||
BC004660.1 | - | 3, 4 | 504 | 3497913 | ||
BY749669.1 | - | 3, 4 | 555 |