Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144789.2 | NP_659038.2 | 3 | 275 | ||
GenBank mRNA | AK144820.1 | - | 3 | 180 | ||
BC072597.1 | - | 2 | 113 |