Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144789.2 | NP_659038.2 | 24 | 3281 | ||
GenBank mRNA | AK053449.1 | - | 2 | 336 | ||
AK144820.1 | - | 24 | 3186 | |||
AY063457.1 | - | 5 | 839 | |||
BC046433.1 | - | 5 | 840 | |||
BC072597.1 | - | 22 | 2968 |