Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly6g6d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033478.2 | NP_258439.1 | 1 | 15 | ||
GenBank mRNA | AJ315548.1 | - | 1 | 15 | ||
AJ315549.1 | - | 1 | 15 | |||
BC131988.1 | - | 1 | 51 | 40130265 | ||
BC131990.1 | - | 1 | 51 | 40130267 |