Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009620.1 | NP_033750.1 | 1 | 921 | ||
GenBank mRNA | AK076647.1 | - | 1 | 982 | ||
AY158688.1 | - | 1 | 921 | |||
BC150159.1 | - | 1 | 925 | 40141514 | ||
BC150481.1 | - | 1 | 925 | 40141515 |