Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adora2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009630.3 | NP_033760.2 | 3 | 1076 | ||
GenBank mRNA | AK037787.1 | - | 2 | 298 | ||
BC110692.1 | - | 2 | 576 | 30242398 | ||
U05672.1 | - | 2 | 800 | |||
Y13346.1 | - | 2 | 561 |
Set Membership: |
G-protein coupled receptor |