Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007414.3 | NP_031440.1 | 4 | 1202 | ||
GenBank mRNA | AF244347.1 | - | 1171 | |||
AK003751.1 | - | 1 | 285 | |||
AK010501.1 | - | 4 | 1167 | |||
AK146280.1 | - | 4 | 1172 | |||
AK153498.1 | - | 4 | 1167 | |||
AK171798.1 | - | 4 | 1172 | |||
BC003437.1 | - | 4 | 1160 | 2647247 | ||
BC058232.1 | - | 4 | 1159 | |||
L13290.1 | - | 3 | 999 |