Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adssl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007421.2 | NP_031447.1 | 12 | 1271 | ||
GenBank mRNA | AK089520.1 | - | 12 | 1246 | ||
AK146907.1 | - | 12 | 1274 | |||
AK149634.1 | - | 12 | 1246 | |||
AK150874.1 | - | 12 | 1242 | |||
BC039943.1 | - | 12 | 1302 | 4014672 | ||
M74495.1 | - | 12 | 1229 |