Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286062.1 | NP_001272991.1 | 9 | 1883 | ||
NM_009640.4 | NP_033770.2 | 9 | 1886 | |||
GenBank mRNA | AK088439.1 | - | 9 | 1887 | ||
AK172868.1 | - | 9 | 1703 | |||
BC067410.1 | - | 9 | 1704 | 30436666 | ||
U83509.1 | - | 9 | 1676 |