Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102446.1 | NP_001095916.1 | 2 | 231 | ||
NM_009653.3 | NP_033783.1 | 2 | 231 | |||
GenBank mRNA | AK002642.1 | - | 2 | 202 | ||
AK077610.1 | - | 2 | 203 | |||
AK165257.1 | - | 2 | 194 | |||
AK165262.1 | - | 2 | 205 | |||
AK172094.1 | - | 2 | 195 | |||
BC150870.1 | - | 2 | 178 | 9087781 | ||
BY035435.1 | - | 2 | 234 | |||
M15268.1 | - | 2 | 189 | |||
M63244.1 | - | 2 | 201 |