Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163266.1 | NP_001156738.1 | 2 | 280 | ||
NM_138721.2 | NP_620046.1 | 2 | 382 | |||
GenBank mRNA | AK143443.1 | - | 2 | 321 | ||
BC024543.1 | - | 2 | 283 | 4989202 | ||
BC026476.1 | - | 2 | 311 | 4489522 |