Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177307.1 | NP_001170778.1 | 6 | 816 | ||
NM_001177308.1 | NP_001170779.1 | 5 | 689 | |||
NM_007438.4 | NP_031464.1 | 5 | 739 | |||
GenBank mRNA | AK002561.1 | - | 5 | 628 | ||
AK011238.1 | - | 1 | 149 | |||
AK145639.1 | - | 5 | 626 | |||
AK154295.1 | - | 5 | 622 | |||
AK162290.1 | - | 5 | 625 | |||
BC043026.1 | - | 5 | 615 | 5691975 | ||
BC050896.1 | - | 5 | 657 | 5715436 | ||
BC066218.1 | - | 5 | 635 | 30054842 | ||
BC066801.1 | - | 5 | 635 | 30072521 | ||
BC089495.1 | - | 5 | 668 | 30312668 | ||
CF617557.1 | - | 5 | 575 | |||
EF662059.1 | - | 5 | 670 | |||
EF662060.1 | - | 5 | 620 | |||
Y00516.1 | - | 5 | 585 |