Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009658.3 | NP_033788.3 | 8 | 852 | ||
GenBank mRNA | AK028079.1 | - | 8 | 807 | ||
AK138000.1 | - | 5 | 1842 | |||
AK139250.1 | - | 8 | 807 | |||
AK149861.1 | - | 8 | 842 | |||
AK150197.1 | - | 8 | 807 | |||
AK153949.1 | - | 8 | 836 | |||
AK164682.1 | - | 8 | 807 | |||
AK170663.1 | - | 8 | 854 | |||
AK193164.1 | - | 1 | 18 | |||
BC004725.1 | - | 8 | 799 | 3582788 | ||
BC021655.1 | - | 8 | 845 | 3708927 | ||
BC085310.1 | - | 8 | 784 | 6485416 | ||
D32250.1 | - | 8 | 793 | |||
L39795.1 | - | 8 | 784 | |||
U29152.1 | - | 8 | 803 |