Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081103.2 | NP_001074572.2 | 4 | 808 | ||
GenBank mRNA | AF328907.1 | - | 3 | 204 | ||
AK080310.1 | - | 4 | 766 | |||
AK173183.1 | - | 4 | 808 | |||
AM712359.1 | - | 4 | 447 | |||
BC137881.1 | - | 4 | 714 | 8860900 | ||
BC145001.1 | - | 4 | 716 | 9053549 |