Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009662.2 | NP_033792.1 | 9 | 1398 | ||
GenBank mRNA | AK089344.1 | - | 9 | 1352 | ||
AK136868.1 | - | 9 | 1367 | |||
AK137481.1 | - | 9 | 1356 | |||
AK169938.1 | - | 9 | 1355 | |||
AK170016.1 | - | 9 | 1255 | |||
AK170434.1 | - | 9 | 1355 | |||
AK170879.1 | - | 9 | 1354 | |||
AK171191.1 | - | 9 | 1347 | |||
AK171413.1 | - | 9 | 1355 | |||
BC139102.1 | - | 9 | 1325 | 8862124 | ||
BC141213.1 | - | 9 | 1325 | 9055825 | ||
L42198.1 | - | 9 | 1244 |