Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054090.1 | NP_473431.1 | 1 | 804 | ||
GenBank mRNA | AB061228.1 | - | 1 | 804 |