Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pop5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026398.4 | NP_080674.1 | 4 | 505 | ||
GenBank mRNA | AK005290.1 | - | 3 | 261 | ||
AK012421.1 | - | 3 | 254 | |||
AK012523.1 | - | 263 | ||||
AK013257.1 | - | 2 | 242 | |||
BC022670.1 | - | 4 | 529 | 4224973 |