Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tirap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177845.1 | NP_001171316.1 | 5, 6 | 565 | ||
NM_001177846.1 | NP_001171317.1 | 5, 6 | 547 | |||
NM_001177847.1 | NP_001171318.1 | 5, 6 | 493 | |||
NM_054096.2 | NP_473437.2 | 5, 6 | 557 | |||
GenBank mRNA | AF378130.1 | - | 5, 6 | 466 | ||
AF410784.1 | - | 4, 5 | 389 | |||
AK036431.1 | - | 5, 6 | 486 | |||
AK080135.1 | - | 5, 6 | 566 | |||
AK151117.1 | - | 5, 6 | 495 | |||
AK168494.1 | - | 5, 6 | 485 | |||
BC005584.1 | - | 2, 3 | 223 | 3157913 | ||
BY740695.1 | - | 5, 6 | 553 | |||
BY743490.1 | - | 5, 6 | 493 |