Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mat1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133653.3 | NP_598414.1 | 7 | 1090 | ||
GenBank mRNA | AK149542.1 | - | 6 | 841 | ||
AK153832.1 | - | 6 | 844 | |||
BC011211.1 | - | 6 | 828 | 4196928 | ||
L13622.1 | - | 6 | 842 |