Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054103.1 | NP_473444.1 | 7 | 1135 | ||
GenBank mRNA | AK014819.1 | - | 7 | 1135 | ||
AM056057.1 | - | 4 | 403 | |||
BC117798.1 | - | 4 | 424 | |||
BC117799.1 | - | 4 | 424 | |||
BC127158.1 | - | 4 | 424 |