Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009670.4 | NP_033800.2 | 6 | 1320 | ||
NM_146005.3 | NP_666117.2 | 29 | 3791 | |||
NM_170687.3 | NP_733788.2 | 6 | 1320 | |||
NM_170688.2 | NP_733789.1 | 29 | 3791 | |||
NM_170689.2 | NP_733790.2 | 28 | 3737 | |||
NM_170690.2 | NP_733791.2 | 28 | 3728 | |||
NM_170728.2 | NP_733924.2 | 27 | 3674 | |||
NM_170729.2 | NP_733925.2 | 27 | 3674 | |||
NM_170730.2 | NP_733926.2 | 4 | 1010 | |||
GenBank mRNA | AF212924.1 | - | 4 | 510 | ||
AK035681.1 | - | 6 | 1048 | |||
AK141720.1 | - | 29 | 3646 | |||
AK157735.1 | - | 7 | 1068 | |||
AK162007.1 | - | 6 | 978 | |||
BC021657.1 | - | 27 | 3544 | 4188590 | ||
BC026152.1 | - | 6 | 921 | |||
L40631.1 | - | 6 | 1323 | |||
L40632.1 | - | 29 | 3333 | |||
U89274.1 | - | 8 | 937 | |||
U89275.1 | - | 8 | 937 |