Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankfy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009671.5 | NP_033801.4 | 6 | 731 | ||
GenBank mRNA | AB011370.2 | - | 6 | 729 | ||
AB098329.1 | - | 6 | 729 | |||
BC139231.1 | - | 6 | 742 | 8862253 | ||
BC145583.1 | - | 6 | 742 | 9054149 |