Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013471.2 | NP_038499.2 | 4 | 229 | ||
GenBank mRNA | AK032703.1 | - | 3 | 257 | ||
AK132293.1 | - | 5 | 271 | |||
AK150486.1 | - | 4 | 190 | |||
AK150614.1 | - | 4 | 176 | |||
AK151054.1 | - | 4 | 176 | |||
AK151236.1 | - | 4 | 208 | |||
AK167338.1 | - | 4 | 230 | |||
AK168390.1 | - | 4 | 201 | |||
AK168487.1 | - | 4 | 210 | |||
AK168917.1 | - | 3 | 234 | |||
AK170447.1 | - | 4 | 175 | |||
BC055871.1 | - | 4 | 189 | 4947415 | ||
U72941.1 | - | 4 | 200 | |||
U95371.1 | - | 4 | 200 |