Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013471.2 | NP_038499.2 | 7 | 560 | ||
GenBank mRNA | AK032703.1 | - | 6 | 588 | ||
AK132293.1 | - | 8 | 602 | |||
AK150486.1 | - | 7 | 521 | |||
AK150614.1 | - | 7 | 507 | |||
AK151054.1 | - | 7 | 507 | |||
AK151236.1 | - | 7 | 539 | |||
AK167338.1 | - | 7 | 561 | |||
AK168390.1 | - | 7 | 532 | |||
AK168487.1 | - | 7 | 541 | |||
AK168917.1 | - | 6 | 565 | |||
AK170447.1 | - | 7 | 506 | |||
BC055871.1 | - | 7 | 520 | 4947415 | ||
U72941.1 | - | 7 | 531 | |||
U95371.1 | - | 7 | 531 |