Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap4s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021710.3 | NP_068356.1 | 6 | 786 | ||
GenBank mRNA | AF092093.1 | - | 6 | 715 | ||
AK005283.1 | - | 6 | 536 | |||
AK178071.1 | - | 5 | 334 | |||
BC053339.1 | - | 6 | 546 | 5370721 | ||
BP426557.1 | - | 4 | 241 |