Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007462.3 | NP_031488.2 | 7 | 965 | ||
GenBank mRNA | AK045053.1 | - | 7 | 936 | ||
AK046280.1 | - | 7 | 955 | |||
AK080907.1 | - | 7 | 965 | |||
AK082724.1 | - | 7 | 711 | |||
BC141141.1 | - | 6 | 678 | 9055753 | ||
M88127.1 | - | 6 | 654 |